accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119301-59-6

IUPAC Name: 4-[1,2,2-tris(4-hydroxyphenyl)ethenyl]phenol
Synonyms: 4-[1,2,2-tris(4-hydroxyphenyl)ethenyl]phenol
Molecular Weight (g/mol): 396.44
Molecular Formula: C26H20O4
InChi Key: QQUZHNPGWNIYMK-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results