accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119396-88-2

H3CONFOCH3

CAS RN 119396-88-2

IUPAC Name: 2-fluoro-4,5-dimethoxybenzonitrile
Synonyms: 2-fluoro-4,5-dimethoxybenzonitrile
Molecular Weight (g/mol): 181.17
Molecular Formula: C9H8FNO2
InChi Key: QZKGCLSVCWWARO-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C#N)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results