accessibility menu, dialog, popup

UserName

Viewing profile as user:
FN

CAS RN 1194-02-1

IUPAC Name: 4-fluorobenzonitrile
Synonyms: benzonitrile, 4-fluoro-
Molecular Weight (g/mol): 121.11
Molecular Formula: C7H4FN
InChi Key: AEKVBBNGWBBYLL-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C#N
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (2)
  • (4)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (4)
  • (6)
  • (3)
  • (7)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (3)