accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 1194-18-9

IUPAC Name: cycloheptane-1,3-dione
Synonyms: cycloheptane-1,3-dione
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: DBOVMTXPZWVYAQ-UHFFFAOYSA-N
SMILES: O=C1CCCCC(=O)C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results