accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClO

CAS RN 1194-64-5

IUPAC Name: 2-chloro-6-methylbenzaldehyde
Synonyms: 2-chloro-6-methylbenzaldehyde
Molecular Weight (g/mol): 154.59
Molecular Formula: C8H7ClO
InChi Key: CCYFXIJPJFSTSU-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C(Cl)=CC=C1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results