accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CS

CAS RN 1195-14-8

IUPAC Name: 2-methyl-1-benzothiophene
Synonyms: 2-methylbenzo[b]thiophene
Molecular Weight (g/mol): 148.22
Molecular Formula: C9H8S
InChi Key: BLZKSRBAQDZAIX-UHFFFAOYSA-N
SMILES: CC1=CC2=CC=CC=C2S1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)