accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNH

CAS RN 1195-42-2

IUPAC Name: N-(propan-2-yl)cyclohexanamine
Synonyms: cyclohexylamine, N-isopropyl-
Molecular Weight (g/mol): 141.26
Molecular Formula: C9H19N
InChi Key: UYYCVBASZNFFRX-UHFFFAOYSA-N
SMILES: CC(C)NC1CCCCC1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)