accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1195-98-8

IUPAC Name: 2-methyl-2-phenylpropanenitrile
Synonyms: 2-methyl-2-phenylpropanenitrile
Molecular Weight (g/mol): 145.21
Molecular Formula: C10H11N
InChi Key: PGQTYXFMSZUGOW-UHFFFAOYSA-N
SMILES: CC(C)(C#N)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results