accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119516-86-8

IUPAC Name: 4-(2,5-dimethyl-1H-pyrrol-1-yl)benzonitrile
Synonyms: 4-(2,5-dimethylpyrrol-1-yl)benzonitrile
Molecular Weight (g/mol): 196.25
Molecular Formula: C13H12N2
InChi Key: FNDFKQYZEDOHRC-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)N1C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results