accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119586-44-6

IUPAC Name: N-(4-{2-[4'-(2-{4-[bis(4-methylphenyl)amino]phenyl}ethenyl)-[1,1'-biphenyl]-4-yl]ethenyl}phenyl)-4-methyl-N-(4-methylphenyl)aniline
Synonyms: N-(4-{2-[4'-(2-{4-[bis(4-methylphenyl)amino]phenyl}ethenyl)-[1,1'-biphenyl]-4-yl]ethenyl}phenyl)-4-methyl-N-(4-methylphenyl)aniline
Molecular Weight (g/mol): 749.01
Molecular Formula: C56H48N2
InChi Key: OSQXTXTYKAEHQV-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=C(C=CC2=CC=C(C=C2)C2=CC=C(C=CC3=CC=C(C=C3)N(C3=CC=C(C)C=C3)C3=CC=C(C)C=C3)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results