accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNN

CAS RN 1196-57-2

IUPAC Name: 1,2-dihydroquinoxalin-2-one
Synonyms: quinoxaline-2-one
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: FFRYUAVNPBUEIC-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2N=C1
Percent Purity 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)