CAS RN 1197-09-7
CAS RN 1197-09-7
IUPAC Name:
1-(3,4-dihydroxyphenyl)ethan-1-one
Synonyms:
3',4'-dihydroxyacetophenone qingxintong acetopyrocatechol
Molecular Weight (g/mol):
152.15
Molecular Formula:
C8H8O3
InChi Key:
UCQUAMAQHHEXGD-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(O)C(O)=C1
1
–
1
of
1
results
1
3',4'-Dihydroxyacetophenone 98.0+%, TCI America™

CAS: 1197-09-7 Molecular Formula: C8H8O3 Molecular Weight (g/mol): 152.149 MDL Number: MFCD00016439 InChI Key: UCQUAMAQHHEXGD-UHFFFAOYSA-N Synonym: 4-Acetocatechol, 4-Acetylpyrocatechol PubChem CID: 14530 ChEBI: CHEBI:19868 IUPAC Name: 1-(3,4-dihydroxyphenyl)ethanone SMILES: CC(=O)C1=CC(=C(C=C1)O)O