accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1197-19-9

IUPAC Name: 4-(dimethylamino)benzonitrile
Synonyms: P-cyano-N,N-dimethylaniline
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: JYMNQRQQBJIMCV-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C#N
Boiling Point 
  • (2)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (4)