CAS RN 1197160-28-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PI 3-K/mTOR Inhibitor III, PKI-179
The PI 3-K/mTOR Inhibitor III, PKI-179, also referenced under CAS 1197160-28-3, controls the biological activity of PI 3-K/mTOR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.