accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119793-66-7

IUPAC Name: [(2R)-3-carboxy-2-(propanoyloxy)propyl]trimethylazanium chloride
Synonyms: propionyl-L-carnitine chloride
Molecular Weight (g/mol): 253.72
Molecular Formula: C10H20ClNO4
InChi Key: KTFMPDDJYRFWQE-DDWIOCJRSA-N
SMILES: [Cl-].CCC(=O)O[C@H](CC(O)=O)C[N+](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results