accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1198-14-7

IUPAC Name: 5-bromoquinolin-8-ol
Synonyms: 5-bromo-8-hydroxyquinoline
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: WIIUANWSGSTCLG-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results