accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1198-37-4

IUPAC Name: 2,4-dimethylquinoline
Synonyms: 2,4-dimethylquinoline
Molecular Weight (g/mol): 157.22
Molecular Formula: C11H11N
InChi Key: ZTNANFDSJRRZRJ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C2C=CC=CC2=N1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)