accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FFNH2FF

CAS RN 1198-64-7

IUPAC Name: tetrafluorobenzene-1,4-diamine
Synonyms: tetrafluorobenzene-1,4-diamine
Molecular Weight (g/mol): 180.11
Molecular Formula: C6H4F4N2
InChi Key: FVFYRXJKYAVFSB-UHFFFAOYSA-N
SMILES: NC1=C(F)C(F)=C(N)C(F)=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results