accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119831-72-0

CH3CH3SOONHNOHOHNOONH2CH3OH3CH3C(E/Z)

CAS RN 119831-72-0

IUPAC Name: 5-{[amino(2,2,5,7,8-pentamethyl-3,4-dihydro-2H-1-benzopyran-6-sulfonamido)methylidene]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms: 5-{[amino(2,2,5,7,8-pentamethyl-3,4-dihydro-1-benzopyran-6-sulfonamido)methylidene]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 662.8
Molecular Formula: C35H42N4O7S
InChi Key: QTWZCODKTSUZJN-UHFFFAOYNA-N
SMILES: CC1=C2OC(C)(C)CCC2=C(C)C(=C1C)S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)