accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrH3C

CAS RN 1198396-46-1

IUPAC Name: 9-bromo-7,7-dimethyl-7H-benzo[c]fluorene
Synonyms: 9-bromo-7,7-dimethylbenzo[c]fluorene
Molecular Weight (g/mol): 323.23
Molecular Formula: C19H15Br
InChi Key: SNUKBTHMZLUJKR-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=C3C=CC=CC3=C2C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results