accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119851-28-4

IUPAC Name: 1-[2-chloro-4-(4-chlorophenoxy)phenyl]ethan-1-one
Synonyms: 1-[2-chloro-4-(4-chlorophenoxy)phenyl]ethanone
Molecular Weight (g/mol): 281.13
Molecular Formula: C14H10Cl2O2
InChi Key: BDTJIVUVQRVLLJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(Cl)C=C(OC2=CC=C(Cl)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results