accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOHOOONHH3CCH3

CAS RN 1198791-74-0

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-{[(4-methoxyphenyl)diphenylmethyl]sulfanyl}-2-methylpropanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-{[(4-methoxyphenyl)diphenylmethyl]sulfanyl}-2-methylpropanoic acid
Molecular Weight (g/mol): 629.77
Molecular Formula: C39H35NO5S
InChi Key: FTIUQZHUKYUVEE-LHEWISCISA-N
SMILES: COC1=CC=C(C=C1)C(SCC(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)