accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1199-65-1

IUPAC Name: 1-ethyl-4-(methoxycarbonyl)pyridin-1-ium iodide
Synonyms: 1-ethyl-4-(methoxycarbonyl)pyridin-1-ium iodide
Molecular Weight (g/mol): 293.1
Molecular Formula: C9H12INO2
InChi Key: NGEAJXXGUZQCPN-UHFFFAOYSA-M
SMILES: [I-].CC[N+]1=CC=C(C=C1)C(=O)OC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)