accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120-00-3

IUPAC Name: N-(4-amino-2,5-diethoxyphenyl)benzamide
Synonyms: N-(4-amino-2,5-diethoxyphenyl)benzamide
Molecular Weight (g/mol): 300.36
Molecular Formula: C17H20N2O3
InChi Key: CNXZLZNEIYFZGU-UHFFFAOYSA-N
SMILES: CCOC1=CC(NC(=O)C2=CC=CC=C2)=C(OCC)C=C1N
Quantity 
  • (2)
  • (3)
Color 
  • (4)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results