accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOSOO

CAS RN 120-18-3

IUPAC Name: naphthalene-2-sulfonic acid
Synonyms: 2-naphthalenesulfonic acid kyselina 2-naftalensulfonova propoxyphene napsylate anhydrous
Molecular Weight (g/mol): 208.23
Molecular Formula: C10H8O3S
InChi Key: KVBGVZZKJNLNJU-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)