accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3

CAS RN 120-25-2

IUPAC Name: 4-ethoxy-3-methoxybenzaldehyde
Synonyms: ethyl-vanillin
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: BERFDQAMXIBOHM-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=O)C=C1OC
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)