accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120-35-4

IUPAC Name: 3-amino-4-methoxy-N-phenylbenzamide
Synonyms: 3-amino-4-methoxy-N-phenylbenzamide
Molecular Weight (g/mol): 242.28
Molecular Formula: C14H14N2O2
InChi Key: LHMQDVIHBXWNII-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1N)C(=O)NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results