accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClClHOO

CAS RN 120-36-5

IUPAC Name: 2-(2,4-dichlorophenoxy)propanoic acid
Synonyms: (+,-)-dichlorprop tri-cornox sprcial dichlorprop, (R)-isomer polyclene dichlorprop, (S)-isomer caswell no. 320 kildip (s)-isomer dichlorprop hedonal DP dichlorprop
Molecular Weight (g/mol): 235.06
Molecular Formula: C9H8Cl2O3
InChi Key: MZHCENGPTKEIGP-UHFFFAOYNA-N
SMILES: CC(OC1=CC=C(Cl)C=C1Cl)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)