accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 120-47-8

IUPAC Name: ethyl 4-hydroxybenzoate
Synonyms: ethylparaben sobrol A mekkings E caswell no. 447 mycocten catalase easeptol aseptin A ethyl parahydroxybenzoate tegosept E
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: NUVBSKCKDOMJSU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (8)
  • (9)
Percent Purity 
  • (4)
  • (5)
  • (10)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (4)
  • (4)
  • (3)
  • (3)
Physical Form 
  • (8)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (5)
  • (2)
  • (3)
  • (5)
  • (1)
  • (4)
  • (7)
  • (5)
  • (3)
  • (8)
  • (9)
  • (4)
  • (5)
  • (10)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (4)
  • (3)
  • (3)
  • (8)
  • (5)
  • (2)
  • (5)
  • (1)