accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNHNO(Z)(E)

CAS RN 120-70-7

IUPAC Name: (6Z)-6-({[3-({[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)propyl]amino}methylidene)cyclohexa-2,4-dien-1-one
Synonyms: (6Z)-6-({[3-({[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)propyl]amino}methylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 282.34
Molecular Formula: C17H18N2O2
InChi Key: BHBZPGSHCVSMEB-PMJBJKLVSA-N
SMILES: O=C1C=CC=C\C1=C\NCCCN\C=C1/C=CC=CC1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)