accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120-71-8

IUPAC Name: 2-methoxy-5-methylaniline
Synonyms: para-cresidine
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: WXWCDTXEKCVRRO-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results