accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO

CAS RN 120-74-1

IUPAC Name: bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Synonyms: bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: FYGUSUBEMUKACF-UHFFFAOYNA-N
SMILES: OC(=O)C1CC2CC1C=C2
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)