accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120-80-9

IUPAC Name: benzene-1,2-diol
Synonyms: catechol phthalic alcohol CI oxidation base 26 o-hydroquinone pyrocatechol (catechol) durafur developer C catechol, 14C-labeled cpd pyrocatechol (product) 2-benzenediol 1 α-hydroxyphenol
Molecular Weight (g/mol): 110.11
Molecular Formula: C6H6O2
InChi Key: YCIMNLLNPGFGHC-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (12)
  • (6)
Boiling Point 
  • (6)
  • (2)
  • (5)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (4)
  • (2)
  • (5)
Physical Form 
  • (5)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (1)
  • (6)
  • (1)
  • (4)
  • (7)
  • (12)
  • (6)
  • (6)
  • (2)
  • (5)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (5)
  • (5)
  • (4)
  • (1)
  • (1)