accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 120-92-3

IUPAC Name: cyclopentanone
Synonyms: cyclopentanone ketopentamethylene adipic ketone adipinketon dumasin
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: BGTOWKSIORTVQH-UHFFFAOYSA-N
SMILES: O=C1CCCC1
Boiling Point 
  • (3)
  • (4)
  • (5)
  • (4)
Molecular Weight (g/mol) 
  • (15)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (5)
  • (3)
  • (4)
Physical Form 
  • (7)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (5)
  • (1)
  • (5)
  • (1)
  • (3)
  • (7)
  • (3)
  • (4)
  • (5)
  • (4)
  • (15)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (3)
  • (4)
  • (7)
  • (5)
  • (4)
  • (5)
  • (1)