accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CFNH2

CAS RN 1200-27-7

IUPAC Name: 1-(4-fluorophenyl)-2-methylpropan-2-amine
Synonyms: 1-(4-fluorophenyl)-2-methylpropan-2-amine
Molecular Weight (g/mol): 167.23
Molecular Formula: C10H14FN
InChi Key: JITFIYFVPMQJOK-UHFFFAOYSA-N
SMILES: CC(C)(N)CC1=CC=C(F)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results