CAS RN 12003-13-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
595.98
Molecular Formula:
AlCl12Ti3
InChi Key:
KSKPGAGLAZXNEO-UHFFFAOYSA-H
SMILES:
Cl[Al](Cl)Cl.Cl[Ti](Cl)Cl
1
–
1
of
1
results
1
Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5%
CAS: 12003-13-3 Molecular Formula: AlCl12Ti3 Molecular Weight (g/mol): 595.98 MDL Number: MFCD00145412 InChI Key: KSKPGAGLAZXNEO-UHFFFAOYSA-H IUPAC Name: titanium(4+) aluminium(3+) heptachloride SMILES: Cl[Al](Cl)Cl.Cl[Ti](Cl)Cl