CAS RN 12003-65-5
IUPAC Name:
aluminium(3+) lanthanum(3+) trioxidandiide
Synonyms:
aluminium(3+) lanthanum(3+) trioxidandiide
Molecular Weight (g/mol):
213.88
Molecular Formula:
AlLaO3
InChi Key:
BOIGHUSRADNYQR-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[Al+3].[La+3]
1
–
3
of
3
results
1
Promotions Available
Lanthanum Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, 100 Orientation, Thermo Scientific Chemicals
Lanthanum Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, 100 Orientation, Thermo Scientific Chemicals
CAS: 12003-65-5 Molecular Formula: LaAlO3 MDL Number: MFCD00144550
2
Promotions Available
Lanthanum aluminum oxide, 99.9% (metals basis)
Lanthanum aluminum oxide, 99.9% (metals basis)
CAS: 12003-65-5 MDL Number: MFCD00144550 Synonym: Lanthanum aluminate
3
Lanthanum aluminum oxide substrate, 10x10x1mm, polished one side, 100 orientation
CAS: 12003-65-5 Molecular Formula: LaAlO3 MDL Number: MFCD00144550