accessibility menu, dialog, popup

UserName

Viewing profile as user:
AlTiTiTi

CAS RN 12003-96-2

IUPAC Name: aluminium(3+) trititanium
Synonyms: aluminium(3+) trititanium
Molecular Weight (g/mol): 170.58
Molecular Formula: AlTi3
InChi Key: JRDVYNLVMWVSFK-UHFFFAOYSA-N
SMILES: [Al+3].[Ti].[Ti].[Ti]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results