accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOAlAlSrSr

CAS RN 12004-37-4

IUPAC Name: dialuminium(3+) distrontium(2+) pentaoxidandiide
Synonyms: strontium aluminate
Molecular Weight (g/mol): 309.2
Molecular Formula: Al2O5Sr2
InChi Key: FNWBQFMGIFLWII-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Sr++].[Sr++]
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)