accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12005-86-6

IUPAC Name: λ⁵-arsenic(5+) sodium hexafluoride
Synonyms: λ⁵-arsenic(5+) sodium hexafluoride
Molecular Weight (g/mol): 211.9
Molecular Formula: AsF6Na
InChi Key: IYPUMRHORYWLBJ-UHFFFAOYSA-H
SMILES: [F-].[F-].[F-].[F-].[F-].[F-].[Na+].[As+5]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results