accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeFeAs

CAS RN 12005-88-8

IUPAC Name: ferrio-λ²-arsanyl iron
Synonyms: ferrio-λ²-arsanyl iron
Molecular Weight (g/mol): 186.61
Molecular Formula: AsFe2
InChi Key: FDKAYGUKROYPRO-UHFFFAOYSA-N
SMILES: [Fe].[Fe]=[As]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results