accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120068-37-3

NH2NNNSOFFFClClFFF(?)

CAS RN 120068-37-3

IUPAC Name: 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-trifluoromethanesulfinyl-1H-pyrazole-3-carbonitrile
Synonyms: fipronil regent C12H4cl2F6N4OS
Molecular Weight (g/mol): 437.14
Molecular Formula: C12H4Cl2F6N4OS
InChi Key: ZOCSXAVNDGMNBV-UHFFFAOYNA-N
SMILES: NC1=C(C(=NN1C1=C(Cl)C=C(C=C1Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)