accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12007-09-9

IUPAC Name: tungstenioborane
Synonyms: tungstenioborane
Molecular Weight (g/mol): 194.65
Molecular Formula: BW
InChi Key: MGRQFMHNBKEJQB-UHFFFAOYSA-N
SMILES: [B+]=[W-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results