accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12007-16-8

IUPAC Name: diboron chromium
Synonyms: diboron chromium
Molecular Weight (g/mol): 73.62
Molecular Formula: B2Cr
InChi Key: UHPOHYZTPBGPKO-UHFFFAOYSA-N
SMILES: [B].[B].[Cr]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results