accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120075-24-3

IUPAC Name: 5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms: 5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 608.71
Molecular Formula: C31H36N4O7S
InChi Key: LKGHIEITYHYVED-UHFFFAOYNA-N
SMILES: COC1=C(C)C(C)=C(C(C)=C1)S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)