accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12009-21-1

IUPAC Name: zirconium(4+) barium(2+) trioxidandiide
Synonyms: zirconium(4+) barium(2+) trioxidandiide
Molecular Weight (g/mol): 276.55
Molecular Formula: BaO3Zr
InChi Key: QLFKDQYBEKOEOT-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Zr+4].[Ba++]
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)