accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNHOO

CAS RN 1201-24-7

IUPAC Name: 5-methyl-1H-indazole-3-carboxylic acid
Synonyms: 5-methyl-1H-indazole-3-carboxylic acid
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: QRTAIBBOZNHRMI-UHFFFAOYSA-N
SMILES: CC1=CC=C2NN=C(C(O)=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results