accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120107-85-9

IUPAC Name: 3-[(1H-imidazol-1-yl)methyl]aniline
Synonyms: 3-(imidazol-1-ylmethyl)aniline
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: HOZXQBSHRUITCX-UHFFFAOYSA-N
SMILES: NC1=CC=CC(CN2C=CN=C2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results