accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12013-10-4

IUPAC Name: disulfanylidenecobalt
Synonyms: disulfanylidenecobalt
Molecular Weight (g/mol): 123.05
Molecular Formula: CoS2
InChi Key: XUKVMZJGMBEQDE-UHFFFAOYSA-N
SMILES: S=[Co]=S
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)