accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12013-47-7

IUPAC Name: calcium oxozirconiumbis(olate)
Synonyms: calcium oxozirconiumbis(olate)
Molecular Weight (g/mol): 179.3
Molecular Formula: CaO3Zr
InChi Key: BJJVDFHQHSBAFC-UHFFFAOYSA-N
SMILES: [Ca++].[O-][Zr]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)